Learn More
Honeywell™ Uvex™ Astrospec 3000™ CB (Custom Bridge) Safety Glasses
Catalog No. 19076070
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-076070 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-076070 Supplier Honeywell™ Supplier No. S2200C
This item is not returnable. View return policy
Custom fit for comfortable protection
Designed specifically for wearers with low-profile nasal area or large-profile cheekbone area, Astrospec 3000 CB spectacles offer features of Astrospec 3000 spectacles plus a customized lens and nose bridge.
Spectacles are lightweight and impact-resistant, with polycarbonate sideshields and browguard for protection against UV, dust, and flying particles. Black frames available with stratch-resistant Ultradura™ coating or Uvextreme™ (XTR) coating for optimal protection from harmful UV light, scratches, fogging, and static.
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Compliance
Both lens coatings meet ANSI Z87.1+ standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1+ standards.
Lens Tint Clear
Frame Color Black