Learn More
ORS Nasco UVEX AVATAR BLK/TEAL CLR HSAF
Supplier:  ORS Nasco 763S2880HS
Encompass_Preferred
Honeywell part# S2880HS. Honeywell Uvex Avatar Safety Glasses Frame Color: Teal Lens Tint: Clear Lens Coating: Hydroshield AF Lens Type: Anti-fog UV Protection: U6 grade Certifications/Compliance: ANSI Z.87.1+CSA Z94.3 Recommended Applications: Safety Temple Color: Black Temple Style: Bayonette
Catalog No. 50-255-1581