Learn More
Honeywell™ Uvex™ Bandit™ Safety Glasses Replacement Lenses
Catalog No. 19003338D
Change view
Click to view available options
Lens Tint:
Amber UD
Clear UD
Clear XTR
Espresso UD
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-003-338D Espresso UD
19-003-338B Clear XTR
19-003-338A Clear UD
19-003-338C Amber UD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-003-338D Supplier Honeywell™ Supplier No. S6312
Contemporary styling, sporty wrap-around temples provide non-slip fit and comfort. Pack of 10 single lenses for use with easy lens replacement system.
  • Sleek, sporty design features a single polycarbonate lens with sideshields and browguards molded into a lightweight, wraparound frame.
  • High-visibility lens is available in a variety of tints, and the patented lens replacement system enables quick, easy lens changes to meet any work application.
  • Lens absorbs 99.9% of harmful UV light and is coated with scratch-resistant Ultradura™ (UD) hardcoat.
  • Clear lenses are also available with exclusive Uvextreme™ AF (XTR) antifog coating that provides additional UV protection and minimizes static and scratching.
TRUSTED_SUSTAINABILITY
Specifications
Lens Type UD
Description Replacement lens
Lens Tint Espresso UD
Product Type Replacement lens
For Use With (Equipment) Bandit Spectacles
No. per Pack 200/Pk.