Learn More
Honeywell™ Uvex™ Bandit™ Safety Glasses Replacement Lenses
Description
- Sleek, sporty design features a single polycarbonate lens with sideshields and browguards molded into a lightweight, wraparound frame.
- High-visibility lens is available in a variety of tints, and the patented lens replacement system enables quick, easy lens changes to meet any work application.
- Lens absorbs 99.9% of harmful UV light and is coated with scratch-resistant Ultradura™ (UD) hardcoat.
- Clear lenses are also available with exclusive Uvextreme™ AF (XTR) antifog coating that provides additional UV protection and minimizes static and scratching.
Specifications
Specifications
| Lens Type | UD |
| Description | Replacement lens |
| Lens Tint | Mirror UD |
| Product Type | Replacement lens |
| For Use With (Equipment) | Bandit Spectacles |
| No. per Pack | 200/Pk. |