These safety goggles are designed for maximum protection and comfort, featuring a lightweight TPE body and a snug fit. They are equipped with clear polycarbonate lenses that have an anti-fog and scratch-resistant coating, ensuring clear vision even in hot and humid conditions. The direct vent design and universal size allow them to fit comfortably over most prescription eyewear, making them a versatile choice for various users and environments.
- Lightweight TPE body provides exceptional comfort.
- Snug fit ensures maximum protection.
- Clear polycarbonate lens offers anti-fog and scratch-resistant properties.
- Direct vent design minimizes fogging in hot and humid conditions.
- Fits comfortably over most prescription eyewear.
- Provides protection against dust and splash.
- Offers UV protection.
- Meets ANSI Z87.1+ standards.