Learn More
MSC Uvex by Honeywell Safety Glasses, OTG, Black Frame, Shade 3.0 Lens, Anti-Scratch/Hard Coat
Catalog No. 5000507976
Encompass
Uvex Astrospec OTG 3001 is a lightweight protective eyewear with a panoramic lens. It is designed to fit comfortably over most prescription eyewear, featuring an economical lens replacement system and including a lifetime frame guarantee.
  • Lightweight protective eyewear with panoramic vision
  • Adjustable lens inclination and temple length
  • Fits comfortably over most prescription eyewear
  • Economical lens replacement system
  • Lifetime frame guarantee
  • Provides 99.9% UVB/UVA protection
  • Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards
  • Polycarbonate lens with Ultra-dura hardcoat and Uvextreme anti-fog coatings
Catalog No. 50-005-07976 Supplier MSC Supplier No. 06519334
Uvex Astrospec OTG 3001 is a lightweight protective eyewear with a panoramic lens. It is designed to fit comfortably over most prescription eyewear, featuring an economical lens replacement system and including a lifetime frame guarantee.
  • Lightweight protective eyewear with panoramic vision
  • Adjustable lens inclination and temple length
  • Fits comfortably over most prescription eyewear
  • Economical lens replacement system
  • Lifetime frame guarantee
  • Provides 99.9% UVB/UVA protection
  • Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards
  • Polycarbonate lens with Ultra-dura hardcoat and Uvextreme anti-fog coatings