Learn More
MSC Uvex by Honeywell Safety Glasses, Smoke Brown Frame, SCT-Reflect 50 Lens, Hardcoat, ANSI Z87.1
Catalog No. 5000508093
Encompass
Uvex Hypershock Safety Glasses feature a lightweight, wraparound full frame in smoke brown and SCT-Reflect 50 polycarbonate lenses with an anti-scratch hardcoat. They provide 99.9% UVB/UVA protection and meet ANSI Z87.1 standards, suitable for various indoor and outdoor work environments. The design prioritizes comfort with a molded nosepiece and temple inserts, offering superior fit, coverage, and peripheral vision.
  • Polycarbonate lens construction.
  • Soft, molded nosepiece and temple inserts for comfort.
  • Wraparound full frame design for high impact protection.
  • Streamlined temples for a secure fit.
  • 100% dielectric, suitable for electrical environments.
  • Provides 99.9% UVB/UVA protection.
  • Certified to meet ANSI Z87.1 standard.
  • Anti-scratch hardcoat on lenses.
  • Ideal for indoor and outdoor use.
  • Integrated hinge mechanisms with dual cam action.
Catalog No. 50-005-08093 Supplier MSC Supplier No. 52883162
Uvex Hypershock Safety Glasses feature a lightweight, wraparound full frame in smoke brown and SCT-Reflect 50 polycarbonate lenses with an anti-scratch hardcoat. They provide 99.9% UVB/UVA protection and meet ANSI Z87.1 standards, suitable for various indoor and outdoor work environments. The design prioritizes comfort with a molded nosepiece and temple inserts, offering superior fit, coverage, and peripheral vision.
  • Polycarbonate lens construction.
  • Soft, molded nosepiece and temple inserts for comfort.
  • Wraparound full frame design for high impact protection.
  • Streamlined temples for a secure fit.
  • 100% dielectric, suitable for electrical environments.
  • Provides 99.9% UVB/UVA protection.
  • Certified to meet ANSI Z87.1 standard.
  • Anti-scratch hardcoat on lenses.
  • Ideal for indoor and outdoor use.
  • Integrated hinge mechanisms with dual cam action.