Learn More
Honeywell™ Uvex™ Coolspec™ 3500 Safety Glasses
Catalog No. 19037551
Change view
Click to view available options
Lens Tint: Gray; Frame Color: Black Frame
Lens Tint: Gray; Frame Color: Gold Frame
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-037-551 Lens Tint: Gray; Frame Color: Black Frame
19-037-556 Lens Tint: Gray; Frame Color: Gold Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-037-551 Supplier Honeywell™ Supplier No. S2351
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Lightweight metal frame for wraparound protection
Lightweight but rugged, single-brow wraparound frame is constructed with fewer solder joints for superior impact resistance. Flexible temples provide increased comfort behind the ears. With contemporary dual-lens styling, single-piece hardcoated polycarbonate lens features a universal nose bridge and integrated sideshield.
Available in a choice of frame colors. Scratch-resistant lens provides up to 99.9% absorption of harmful UV light.
Compliance
All spectacles meet both ANSI Z87.1-1989 and CSA Z94.3-1992 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance All spectacles meet both ANSI Z87.1-1989 and CSA Z94.3-1992 standards.
Lens Tint Gray
Frame Color Black
Lens Material Polycarbonate