Learn More
Honeywell™ Uvex™ Coolspec™ 3500 Safety Glasses
Catalog No. 19037556
Change view
Click to view available options
Lens Tint: Gray; Frame Color: Black Frame
Lens Tint: Gray; Frame Color: Gold Frame
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-037-556 Lens Tint: Gray; Frame Color: Gold Frame
19-037-551 Lens Tint: Gray; Frame Color: Black Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-037-556 Supplier Honeywell™ Supplier No. S2371
This item is not returnable. View return policy
Lightweight metal frame for wraparound protection
Lightweight but rugged, single-brow wraparound frame is constructed with fewer solder joints for superior impact resistance. Flexible temples provide increased comfort behind the ears. With contemporary dual-lens styling, single-piece hardcoated polycarbonate lens features a universal nose bridge and integrated sideshield.
Available in a choice of frame colors. Scratch-resistant lens provides up to 99.9% absorption of harmful UV light.
Compliance
All spectacles meet both ANSI Z87.1-1989 and CSA Z94.3-1992 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance All spectacles meet both ANSI Z87.1-1989 and CSA Z94.3-1992 standards.
Lens Tint Gray
Frame Color Gold
Lens Material Polycarbonate