Learn More
Honeywell™ Uvex™ Falcon Replacement Lenses
Catalog No. 19055457
Change view
Click to view available options
Lens Tint:
Amber
Espresso
SCT-Reflect 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-055-457 SCT-Reflect 50
19-055-453 Espresso
19-055-454 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-055-457 Supplier Honeywell™ Supplier No. S6704
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Sleek, frameless design with wrap-around 9-base unilens. Lightweight comfort and superior coverage providing complete and unobstructed vision.
Lighter than air, the Uvex Falcon soars above the competition as a great value in protective eyewear. The Uvex Falcon's frameless and optically perfected dual nine-base wrap-around lens provides superior coverage and unobstructed peripheral vision.
  • Sleek, frameless design with wrap-around 9-base unilens
  • Easy and economical lens replacement system
  • Box of 10 clear replacement lenses with Ultra-dura Anti-scratch Coating
  • Meets ANSI Z87.1-2003 (High Impact) and tested to meet the requirements of the CSA Z94.3 standard
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Ultradura
Description Replacement lens
Lens Tint SCT-Reflect 50
Product Type Replacement lens