Learn More
Honeywell™ Uvex™ Flame-Retardant Goggle Headbands
Catalog No. 190881688
Change view
Click to view available options
Headband
Catalog No.
19-088-1688 Headband
1 options
This item is not returnable. View return policy
Catalog No. 19-088-1688 Supplier Honeywell™ Supplier No. S823
This item is not returnable. View return policy
Replacement headband made of flame resistant fabric for use with the the Uvex™ Stealth or Fury goggles.
  • Uvex Fury: Sleek, frameless design with wrap-around 9-base unilens. Ideal for dust and small particulates. Soft, flame-resistant, replaceable face foam and headband.
  • Uvex Stealth: Futuristic, low profile design for chemical splash and impact. Toric lens provides superior optics and peripheral vision. Quick adjust headband.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Headband Goggle
For Use With (Equipment) Stealth, Spoggle, Fury
Material Flame retardant fabric