Learn More
Honeywell™ Uvex™ Flashback™ Safety Glasses
Catalog No. 189992445
Change view
Click to view available options
Lens Coating:
Impact-resistant
Lens Tint:
Clear
Catalog No. Lens Coating Lens Tint
18-999-2445 Impact-resistant Clear
1 options
This item is not returnable. View return policy
Catalog No. 18-999-2445 Supplier Honeywell™ Supplier No. S4000C
This item is not returnable. View return policy
Exclusive single-lens design offers fast, economical lens replacement, but has the look of dual-lens eyewear. Patented Duoflex™ cushions minimize pressure behind the ears, with temple and lens adjustments for a custom fit. Impact-resistant polycarbonate lens and molded-in sideshields and browguard deliver impact protection. Lenses absorb >99.9% of UV radiation. Clear lenses come in your choice of antiscratch Ultradura™ (UD) hardcoat or antifog, antistatic, antiscratch Uvextreme™ AF (XTR) coating, or with gray lenses in UD coating. Black frames.
Compliance
Spectacles meet ANSI Z87.1-2010 standards and are CSA Z94.3-1999 high-impact approved.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Spectacles meet ANSI Z87.1-2003 standards and are CSA Z94.3-1999 high-impact approved.
Lens Coating Impact-resistant
Description Clear XTR
Lens Tint Clear
Frame Color Black
Lens Material Polycarbonate