Learn More
ORS Nasco UVEX FLEX SEAL OTG GOGGLE NAVY
Catalog No. 502551575
Encompass_Preferred
Honeywell part# S3400HS. Honeywell Uvex Flex Seal Safety Goggles Headband Material: Neoprene Lens Tint: Clear Lens Type: Anti-Fog Certifications/Compliance: ANSI Z87+ (High Impact) ANSI Z.87.1/1989 CA 21199 CSA Z94.3 For Use With: Abrasions Airborne Particulate Biohazard Chemical Impact Splash
Catalog No. 50-255-1575 Supplier ORS Nasco Supplier No. 763S3400HS
Honeywell part# S3400HS. Honeywell Uvex Flex Seal Safety Goggles Headband Material: Neoprene Lens Tint: Clear Lens Type: Anti-Fog Certifications/Compliance: ANSI Z87+ (High Impact) ANSI Z.87.1/1989 CA 21199 CSA Z94.3 For Use With: Abrasions Airborne Particulate Biohazard Chemical Impact Splash