Learn More
Honeywell™ Uvex Flex Seal™ Safety Goggle Replacement Lens
Catalog No. 19085468
Change view
Click to view available options
Polycarbonate
Catalog No.
19-085-468 Polycarbonate
1 options
This item is not returnable. View return policy
Catalog No. 19-085-468 Supplier Honeywell™ Supplier No. S710HS
This item is not returnable. View return policy
Honeywell™ Uvex Flex Seal™ Safety Goggle Replacement Lens with Hydroshield™ AF coating
  • Uvextreme AF lens coating
  • Hydroshield AF coating
  • Compatible with a variety of Honeywell™ Uvex Flex Seal™ safety goggles
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87+ (High Impact), ANSI Z.87.1/1989, CA 21,199, CSA Z94.3
Lens Tint Clear
Product Type Replacement lens
For Use With (Application) S3400HS, S3405HS
No. per Pack 1 Ea.
Disposable Single-use
Lens Material Polycarbonate