Learn More
Uline Ultra-flexible silicone seal Safety Goggles
Catalog No. NC1785805
Encompass
This item has a minimum order quantity of 2 per supplier requirements. Ultra-flexible silicone seal conforms to face. Protects against impact and chemical splash. Indirect vent. HydroShield™ coating guarantees fog-free lenses. 99.9% UV protection.
This item is not returnable. View return policy
Catalog No. NC1785805 Supplier Uline Supplier No. S20752
This item has a minimum order quantity of 2 per supplier requirements. Ultra-flexible silicone seal conforms to face. Protects against impact and chemical splash. Indirect vent. HydroShield™ coating guarantees fog-free lenses. 99.9% UV protection.