Learn More
Honeywell™ Uvex™ Futura™ Chemical Splash Goggles Replacement Lenses
Catalog No. 172755C
Change view
Click to view available options
lens size: Large
Catalog No.
17-275-5C lens size: Large
1 options
Catalog No. 17-275-5C Supplier Honeywell™ Supplier No. S780C
Replacement lenses for the Uvex™ Futura™ are clear lenses with Uvextreme anti-fog coating. Futura goggles have an easy and economical lens replacement system.
Uvex Future features a wrap-around sports style that provides excellent panoramic vision. Frosted brow guard reduces glare. Indirect venting increases air flow to reduce fogging.
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement lens
Lens Tint Clear
Product Type Replacement lens
For Use With (Equipment) Chemical splash goggles