Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Catalog No. 18813398
Change view
Click to view available options
Lens tint: amber; Magnfication: +2.5
Catalog No.
18-813-398 Lens tint: amber; Magnfication: +2.5
1 options
Catalog No. 18-813-398 Supplier Honeywell™ Supplier No. 95891201
Add to Cart
Add to Cart
High-impact protection
  • Soft, flexible nasal fingers
  • Padded temple tips and cushioned browbar
  • Adjustable temple length and ratcheting lens inclination provides for secure fit
  • Easy and economical lens replacement (see Cat. No. 18-813-424)
  • With Ultra-dura™ antiscratch coating
  • Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.