Learn More
Honeywell™ Uvex™ Genesis X2™ Safety Glasses: Replacement Lenses
Catalog No. 17377416
Change view
Click to view available options
For Use With (Application):
Light mirror coating for varying light conditions
Lens Tint:
SCT-Reflect 50
Catalog No. For Use With (Application) Lens Tint
17-377-416 Light mirror coating for varying light conditions SCT-Reflect 50
1 options
Catalog No. 17-377-416 Supplier Honeywell™ Supplier No. S6924
Replacement lenses for Uvex Genesis X2 safety glasses (cat. no. 17377422)
Uvextreme™ Antifog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
Ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
Compliance
Exceed ANSI™ Z87.1-2010 standards. Meet stringent military V0 ballistic impact requirements.
Warranty and Services
Lifetime frame guarantee
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Description Replacement lenses for Uvex Genesis X2 spectacles
Lens Tint SCT-Reflect 50
Product Type Replacement lens
For Use With (Equipment) Uvex Genesis X2 spectacles
For Use With (Application) Light mirror coating for varying light conditions
No. per Pack 10/Pk.