Learn More
Honeywell™ Uvex™ Horizon™ Safety Glasses
Catalog No. 191302473
Change view
Click to view available options
Lens Tint:
Cobalt Blue
IR Filter Shade 5.0
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-130-2473 Cobalt Blue
19-130-2471 IR Filter Shade 5.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-2473 Supplier Honeywell™ Supplier No. S214
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Classic flip-up style offers versatility for workers in unique environments. Shaded lens designed to protect against high intensity light sources, while the clear lens offers impact resistance.
The Uvex Horizon™ is perfect for welders and industrial workers who require a shaded lens while working with high intensity light sources, and then need a traditional clear impact-resistant polycarbonate lens when their task is complete. The Uvex Horizon is lightweight, allowing greater comfort for longer wearing time and is suitable for Rx lenses. Meets ANSI Z87+ (High Impact) standard.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, Scratch-resistant
Lens Tint Cobalt Blue
Frame Color Black
Temple Style Spatula w/metal core
Lens Material Polycarbonate