Learn More
Honeywell™ Uvex™ Horizon™ Safety Glasses
Catalog No. p-3543493
Change view
Click to view available options
Lens Tint:
Cobalt Blue
IR Filter Shade 5.0
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-130-2471 IR Filter Shade 5.0
19-130-2473 Cobalt Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-2471 Supplier Honeywell™ Supplier No. S212
This item is not returnable. View return policy
Classic flip-up style offers versatility for workers in unique environments. Shaded lens designed to protect against high intensity light sources, while the clear lens offers impact resistance.
The Uvex Horizon™ is perfect for welders and industrial workers who require a shaded lens while working with high intensity light sources, and then need a traditional clear impact-resistant polycarbonate lens when their task is complete. The Uvex Horizon is lightweight, allowing greater comfort for longer wearing time and is suitable for Rx lenses. Meets ANSI Z87+ (High Impact) standard.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint IR Filter Shade 5.0
Frame Color Black
Temple Style Spatula w/metal core
Lens Material Polycarbonate