Learn More
Honeywell™ Uvex™ Ignite™ Safety Glasses
Description
The Uvex Ignite delivers exceptional comfort, protection and style in a lightweight, sleek, frameless design. With its dual nine-base, wrap-around unilens, the Uvex Ignite delivers sport-styling with close to the-face coverage for when you need it most. The adjustable nose piece and soft temple tip padstruly make the Uvex Ignite a pleasure to wear. Add to that a bold temple style, hot colors and an array of high-performance lens tints and the Uvex Ignite stands out from the crowd.
- Two bold temple styles available: hot metallic red with silver accents or more traditional black with silver accents
- Unisex design appeals to a broad range of workers
- Soft, saddle pad nosepiece reduces slippage and provides a personalized, comfortable fit
- Soft temple tip pads ensure a fit that is both comfortable and secure
Specifications
Specifications
| Lens Coating | Hardcoat |
| Lens Tint | Silver Mirror |
| Frame Color | Red |
| Temple Style | Bold |
| Frame Type | Frameless design offers 180-degrees of distortion-free, unobstructed vision |