Learn More
Honeywell™ Uvex™ Ignite™ Safety Goggles
Catalog No. 19053950
Change view
Click to view available options
Uvex Ignite Safety Goggles
Catalog No.
19-053-950 Uvex Ignite Safety Goggles
1 options
This item is not returnable. View return policy
Catalog No. 19-053-950 Supplier Honeywell™ Supplier No. S4410X
This item is not returnable. View return policy
Clear lens with temple arms in hot metallic red with silver accents. Sleek wrap-around unilens provides close to the face protection and superior coverage. Lightweight, frameless design offers complete 180°, distortion-free vision. Saddle pad nosepiece reduces slippage.
The Uvex Ignite delivers exceptional comfort, protection and style in a lightweight, sleek, frameless design. The Uvex Ignite delivers sport-styling with close to the-face coverage for when you need it most. The adjustable nose piece and soft temple tip padstruly make the Uvex Ignite a pleasure to wear.
  • Sporty, wrap-around, 9-base unilens with Uvextra Anti-fog coating
  • Unisex design appeals to a broad range of workers
  • Soft, saddle pad nosepiece provides a personalized, comfortable fit
  • Soft temple tip pads ensure a fit that is both comfortable and secure
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety goggles
Frame Color Red, Silver
Includes Saddle pad nosepiece; Soft temple tip pad; Nine base clear unilens
Material Polycarbonate