Learn More
MSC Uvex 880 Series Hard Hat Clip Lenses, Polycarbonate, Welding Shade 8, Universal, 1 ea
Catalog No. 5000507983
Encompass
These clip-on lenses are designed to enhance vision by attaching directly to hard hat or bump cap brims. The full-front style allows users to quickly flip the lenses down for protection or up when not needed, providing a full 180-degree arc of movement to clear the line of vision. Constructed with a stainless steel frame and polycarbonate lenses, these clip-ons offer reliable performance in construction and general-purpose applications.
  • Features a welding shade 8 lens with U6 ultraviolet and R8 infra-red scale protection
  • Includes a scratch-resistant lens coating for durability
  • Attaches easily with an integral spring clip without the need for tools or fasteners
  • Compatible with all hard hats
  • Provides 99.9% UV protection
  • X-ray detectable design
Catalog No. 50-005-07983 Supplier MSC Supplier No. 62309836
These clip-on lenses are designed to enhance vision by attaching directly to hard hat or bump cap brims. The full-front style allows users to quickly flip the lenses down for protection or up when not needed, providing a full 180-degree arc of movement to clear the line of vision. Constructed with a stainless steel frame and polycarbonate lenses, these clip-ons offer reliable performance in construction and general-purpose applications.
  • Features a welding shade 8 lens with U6 ultraviolet and R8 infra-red scale protection
  • Includes a scratch-resistant lens coating for durability
  • Attaches easily with an integral spring clip without the need for tools or fasteners
  • Compatible with all hard hats
  • Provides 99.9% UV protection
  • X-ray detectable design