Learn More
MSC Honeywell Uvex 880 Series Klip Lift, Shade 8, Cobalt Blue, Full Front, 1 Ea.
Catalog No. 5000510009
Encompass
The 880 Series Klip Lift is a full-front welding shade 8 lens designed to provide convenient vision enhancement for industrial applications. This cobalt blue lens is engineered for compatibility with hard hats or bump caps, allowing for seamless integration with existing protective eyewear. It features a versatile flip-up design with a 180° arc of movement, enabling quick transitions between protected and clear viewing during tasks such as welding, cutting, steelmaking, and foundry work.
  • Provides welding shade 8 protection
  • Features a full-front lens design
  • Includes an integral spring clip for secure, tool-free mounting to hard hat or bump cap brims
  • Allows for a 180° arc of movement for rapid flipping
  • Compatible for use in conjunction with existing safety eyewear
  • Suitable for use in steel and metals, manufacturing, mining, and oil and gas industries
Catalog No. 50-005-10009 Supplier MSC Supplier No. 06716609
The 880 Series Klip Lift is a full-front welding shade 8 lens designed to provide convenient vision enhancement for industrial applications. This cobalt blue lens is engineered for compatibility with hard hats or bump caps, allowing for seamless integration with existing protective eyewear. It features a versatile flip-up design with a 180° arc of movement, enabling quick transitions between protected and clear viewing during tasks such as welding, cutting, steelmaking, and foundry work.
  • Provides welding shade 8 protection
  • Features a full-front lens design
  • Includes an integral spring clip for secure, tool-free mounting to hard hat or bump cap brims
  • Allows for a 180° arc of movement for rapid flipping
  • Compatible for use in conjunction with existing safety eyewear
  • Suitable for use in steel and metals, manufacturing, mining, and oil and gas industries