Learn More
MSC Uvex Klip Lifts, Shade 6, Blue, Hard Hat Compatible
Catalog No. 5000507033
Encompass
These eye protection klip lifts are designed to attach to hard hat or bump cap brims, allowing them to be worn with existing protective eyewear. They offer quick vision enhancement with a flip-down mechanism, making them ideal for applications requiring temporary specialized eye protection. An integral spring clip ensures secure attachment without the need for additional tools or fasteners. This model features a full front lens with a shade 6 blue tint.
  • Designed to fit onto hard hat or bump cap brims.
  • Worn in conjunction with existing protective eyewear.
  • Lenses can be quickly flipped down to enhance vision and then flipped back up when not needed.
  • Integral spring clip holds securely without screws, fasteners, or tools.
  • Features a full front lens.
  • Provides shade 6 blue eye protection.
  • Frame constructed from stainless steel.
Catalog No. 50-005-07033 Supplier MSC Supplier No. 72969678
These eye protection klip lifts are designed to attach to hard hat or bump cap brims, allowing them to be worn with existing protective eyewear. They offer quick vision enhancement with a flip-down mechanism, making them ideal for applications requiring temporary specialized eye protection. An integral spring clip ensures secure attachment without the need for additional tools or fasteners. This model features a full front lens with a shade 6 blue tint.
  • Designed to fit onto hard hat or bump cap brims.
  • Worn in conjunction with existing protective eyewear.
  • Lenses can be quickly flipped down to enhance vision and then flipped back up when not needed.
  • Integral spring clip holds securely without screws, fasteners, or tools.
  • Features a full front lens.
  • Provides shade 6 blue eye protection.
  • Frame constructed from stainless steel.