Learn More
Honeywell™ Uvex™ Millennia™ Protective Safety Glasses
Catalog No. 19039229
Change view
Click to view available options
Frame Color:
Black
Blue
Gray
Lens Coating:
Impact-resistant
Impact-resistant, Scratch-resistant
Scratch-resistant
Lens Tint:
Clear
Gray
Silver Mirror
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-039-229 Gray Scratch-resistant Silver Mirror
19-087-375B Black Impact-resistant, Scratch-resistant Gray
19-039-212 Black Scratch-resistant Silver Mirror
19-039-213 Blue Impact-resistant, Scratch-resistant Clear
19-087-375A Black Impact-resistant Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-039-229 missing translation for 'mfr' Honeywell™ missing translation for 'supplierNo' 11150362
This item is not returnable. View return policy
Futuristic wraparound styling with exceptional coverage and protection
  • Lightweight nylon frame with polycarbonate lens
  • Superior side and brow protection
  • High impact dual 9-base wraparound unilens
  • Comfortable universal nose bridge
Order Info
Lenses with Fog-Ban™ coating available for clear vision in humid ambient conditions.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
Lens Coating Scratch-resistant
Description Silver Mirror I/O
Lens Tint Silver Mirror
Frame Color Gray
Lens Material Polycarbonate