Learn More
Honeywell™ Uvex™ Millennia™ Protective Safety Glasses
Catalog No. 19087375A
Change view
Click to view available options
Frame Color:
Black
Blue
Lens Coating:
Impact-resistant
Impact-resistant, Scratch-resistant
Lens Tint:
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-087-375A Black Impact-resistant Clear
19-039-213 Blue Impact-resistant, Scratch-resistant Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-087-375A Supplier Honeywell™ Supplier No. 11150355
This item is not returnable. View return policy
Futuristic wraparound styling with exceptional coverage and protection
  • Lightweight nylon frame with polycarbonate lens
  • Superior side and brow protection
  • High impact dual 9-base wraparound unilens
  • Comfortable universal nose bridge
Order Info
Lenses with Fog-Ban™ coating available for clear vision in humid ambient conditions.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
Lens Coating Impact-resistant
Lens Tint Clear
Frame Color Black
Lens Material Polycarbonate