Learn More
Honeywell™ Uvex™ Millennia™ Protective Safety Glasses
Description
- Lightweight nylon frame with polycarbonate lens
- Superior side and brow protection
- High impact dual 9-base wraparound unilens
- Comfortable universal nose bridge
Order Info
Lenses with Fog-Ban™ coating available for clear vision in humid ambient conditions.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
Specifications
Specifications
| Certifications/Compliance | Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact. |
| Lens Coating | Impact-resistant, Scratch-resistant |
| Lens Tint | Gray |
| Frame Color | Black |
| Lens Material | Polycarbonate |