Learn More
Honeywell™ Uvex™ PrecisionPro™ Eyewear
Description
- Specialized, task-specific SCT-Spectrum Control Technology lens tints optimize lighting and enhance visual perception
- FlexFit wire core temples bend and flex for a customizable, full-comfort fit
- FlexFit nosepiece conforms to virtually any nasal profile and provides a contoured, non-slip fit
- Ratcheting temples enable wearer to adjust lens inclination angle to fit perfectly above cheekbones
- Exceptional peripheral vision and coverage
- Supra-Dura anti-scratch coating makes lenses last longer
Compliance
Exceeds ANSI™ Z87.1-2003.
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | Exceeds ANSI Z87.1-2003. |
| Lens Tint | Clear |
| For Use With (Application) | Maximum visibility for most indoor applications |