Learn More
MSC Uvex Replacement Lens, Genesis Series, Espresso, HydroShield Anti-Fog
Catalog No. 5000508534
Encompass
This replacement lens is designed for use with Genesis protective eyewear. It features an espresso-colored polycarbonate lens with HydroShield Anti-Fog coating, providing enhanced clarity and durability for various work environments.
  • HydroShield Anti-Fog coating provides long-lasting fog-free performance and scratch resistance.
  • Constructed from durable polycarbonate material.
  • Espresso tinted lens for specific light conditions.
  • Designed for use with Genesis protective eyewear.
  • Offers an economical replacement system to extend eyewear life.
  • Meets ANSI Z87.1-2015 (High Impact) and military V0 ballistic impact standards.
  • CSA certified for safety.
Catalog No. 50-005-08534 Supplier MSC Supplier No. 33706334
This replacement lens is designed for use with Genesis protective eyewear. It features an espresso-colored polycarbonate lens with HydroShield Anti-Fog coating, providing enhanced clarity and durability for various work environments.
  • HydroShield Anti-Fog coating provides long-lasting fog-free performance and scratch resistance.
  • Constructed from durable polycarbonate material.
  • Espresso tinted lens for specific light conditions.
  • Designed for use with Genesis protective eyewear.
  • Offers an economical replacement system to extend eyewear life.
  • Meets ANSI Z87.1-2015 (High Impact) and military V0 ballistic impact standards.
  • CSA certified for safety.