Learn More
MSC Uvex Replacement Lens, Gray, Uvextreme Anti-fog
Catalog No. 5000508085
Encompass
This replacement lens features an extended wrap-around design for enhanced cheek and side protection. It is designed with soft, flexible nose fingers that adapt to various nasal profiles and includes adjustable temples and lens inclination for a customized fit. The lens also offers an economical replacement system.
  • Extended wrap-around lens for extra cheek and side coverage (up to 20% more).
  • Rx lens insert shields expensive prescription lenses from damage.
  • Soft, flexible nose fingers conform to fit a variety of nasal profiles.
  • Easy and economical lens replacement system.
  • Adjustable temples and lens inclination for custom fit.
  • Lifetime frame guarantee.
  • Made in USA.
  • Meets ANSI Z87+ (High Impact) regulations.
  • Meets ANSI.Z.87.1/1989, CA 18,819 regulations.
  • Meets CSA Z94.3 standards.
  • Meets Military V0 ballistic test for impact.
Catalog No. 50-005-08085 Supplier MSC Supplier No. 33817479
This replacement lens features an extended wrap-around design for enhanced cheek and side protection. It is designed with soft, flexible nose fingers that adapt to various nasal profiles and includes adjustable temples and lens inclination for a customized fit. The lens also offers an economical replacement system.
  • Extended wrap-around lens for extra cheek and side coverage (up to 20% more).
  • Rx lens insert shields expensive prescription lenses from damage.
  • Soft, flexible nose fingers conform to fit a variety of nasal profiles.
  • Easy and economical lens replacement system.
  • Adjustable temples and lens inclination for custom fit.
  • Lifetime frame guarantee.
  • Made in USA.
  • Meets ANSI Z87+ (High Impact) regulations.
  • Meets ANSI.Z.87.1/1989, CA 18,819 regulations.
  • Meets CSA Z94.3 standards.
  • Meets Military V0 ballistic test for impact.