Learn More
MSC Uvex Replacement Visor, Polycarbonate, Gray, Toric, 2.20 mm
Catalog No. 5000507961
Encompass
This replacement visor is designed for the Uvex Turboshield face shield system. Made from durable polycarbonate, it features a gray, toric-shaped lens that provides excellent optics and an increased field of view. The gray tint minimizes outdoor sunlight and glare, while the visor ensures protection against impact, splash, and debris, meeting ANSI Z87.1-2010 and CSA Z94.3 safety standards.
  • Toric-shaped lens provides excellent optics, increased field of view, and added chin coverage.
  • Gray lens minimizes outdoor sunlight and glare.
  • Constructed from durable polycarbonate material.
  • Uncoated for clear vision.
  • Meets ANSI Z87.1-2010 and CSA Z94.3 safety standards.
Catalog No. 50-005-07961 Supplier MSC Supplier No. 61455531
This replacement visor is designed for the Uvex Turboshield face shield system. Made from durable polycarbonate, it features a gray, toric-shaped lens that provides excellent optics and an increased field of view. The gray tint minimizes outdoor sunlight and glare, while the visor ensures protection against impact, splash, and debris, meeting ANSI Z87.1-2010 and CSA Z94.3 safety standards.
  • Toric-shaped lens provides excellent optics, increased field of view, and added chin coverage.
  • Gray lens minimizes outdoor sunlight and glare.
  • Constructed from durable polycarbonate material.
  • Uncoated for clear vision.
  • Meets ANSI Z87.1-2010 and CSA Z94.3 safety standards.