Learn More
MSC Uvex Replacement Window, Clear, Anti-Fog, Scratch-Resistant, 0.086 in Thick
Catalog No. 5000509473
Encompass
This replacement window is designed for face shields, offering clear vision with anti-fog and scratch-resistant properties. It is built to industry standards, ensuring reliable performance and safety in various work environments.
  • Clear lens for unobstructed vision.
  • Anti-fog coating to maintain clarity in challenging conditions.
  • Scratch-resistant coating for enhanced durability and extended product life.
  • Measures 9 inches high.
  • Has a thickness of 0.086 inches.
  • Compliant with ANSI Z87.1-2010 safety standard.
  • Compliant with CSA Z94.3 safety standard.
Catalog No. 50-005-09473 Supplier MSC Supplier No. 61455549
This replacement window is designed for face shields, offering clear vision with anti-fog and scratch-resistant properties. It is built to industry standards, ensuring reliable performance and safety in various work environments.
  • Clear lens for unobstructed vision.
  • Anti-fog coating to maintain clarity in challenging conditions.
  • Scratch-resistant coating for enhanced durability and extended product life.
  • Measures 9 inches high.
  • Has a thickness of 0.086 inches.
  • Compliant with ANSI Z87.1-2010 safety standard.
  • Compliant with CSA Z94.3 safety standard.