Learn More
MSC Uvex Safety Glasses, Anti-Fog and Anti-Scratch, Polycarbonate, Over-the-Glasses, 1 ea
Catalog No. 5000506692
Encompass
Uvex Avatar Plus OTG 2 Safety Glasses provide protection, durability, and all-day comfort for users wearing prescription eyewear. Engineered with a large frame and field of vision, these glasses feature premium adjustability for a secure fit, making them suitable for various industrial and laboratory applications.
  • Designed to fit over prescription eyewear (OTG)
  • Gray polycarbonate lenses with anti-fog and anti-scratch coating
  • UV protection
  • Semi-frameless design with gray and green frame
  • Adjustable, ratcheting temples
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Features PC unilens with Ultra-dura hardcoat (HC)
Catalog No. 50-005-06692 Supplier MSC Supplier No. 88843925
Uvex Avatar Plus OTG 2 Safety Glasses provide protection, durability, and all-day comfort for users wearing prescription eyewear. Engineered with a large frame and field of vision, these glasses feature premium adjustability for a secure fit, making them suitable for various industrial and laboratory applications.
  • Designed to fit over prescription eyewear (OTG)
  • Gray polycarbonate lenses with anti-fog and anti-scratch coating
  • UV protection
  • Semi-frameless design with gray and green frame
  • Adjustable, ratcheting temples
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Features PC unilens with Ultra-dura hardcoat (HC)