Learn More
MSC Uvex Safety Glasses, Anti-Fog, Polycarbonate, Amber Lenses, Full-Framed
Catalog No. 5000509436
Encompass
These safety glasses offer superior eye protection with a full-framed design. Featuring anti-fog polycarbonate amber lenses, they provide excellent clarity and UV protection, making them suitable for various work environments. The adjustable temples and cushioned browbar ensure a comfortable and secure fit.
  • Anti-fog lens coating for clear vision
  • Polycarbonate lenses for impact resistance
  • Amber lenses enhance contrast and reduce glare
  • Full-framed, wrap-around design for comprehensive coverage
  • Adjustable temples for a customized fit
  • Cushioned browbar for enhanced comfort
  • Meets ANSI Z87.1+ standards for safety
  • Provides UV protection
Catalog No. 50-005-09436 Supplier MSC Supplier No. 41570821
These safety glasses offer superior eye protection with a full-framed design. Featuring anti-fog polycarbonate amber lenses, they provide excellent clarity and UV protection, making them suitable for various work environments. The adjustable temples and cushioned browbar ensure a comfortable and secure fit.
  • Anti-fog lens coating for clear vision
  • Polycarbonate lenses for impact resistance
  • Amber lenses enhance contrast and reduce glare
  • Full-framed, wrap-around design for comprehensive coverage
  • Adjustable temples for a customized fit
  • Cushioned browbar for enhanced comfort
  • Meets ANSI Z87.1+ standards for safety
  • Provides UV protection