Learn More
MSC Uvex Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, Clear Lenses, ANSI Z87.1+
Catalog No. 5000509380
Encompass
These safety glasses feature clear polycarbonate lenses with both anti-fog and scratch-resistant coatings, designed for improved visibility in various conditions. They have a half-framed, wrap-around design with a black and pewter frame, and include adjustable temples for a comfortable and secure fit. The glasses are part of the Genesis Series and meet ANSI Z87.1+ standards, ensuring reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Scratch-resistant lenses enhance durability and longevity.
  • Polycarbonate lenses provide impact resistance for safety.
  • Clear lenses offer unobstructed vision.
  • Half-framed design for a lightweight feel.
  • Adjustable temples ensure a comfortable and secure fit for various head sizes.
  • Meets ANSI Z87.1+ standards for reliable eye protection.
  • Flexible nose bridge for added comfort.
Catalog No. 50-005-09380 Supplier MSC Supplier No. 72055874
These safety glasses feature clear polycarbonate lenses with both anti-fog and scratch-resistant coatings, designed for improved visibility in various conditions. They have a half-framed, wrap-around design with a black and pewter frame, and include adjustable temples for a comfortable and secure fit. The glasses are part of the Genesis Series and meet ANSI Z87.1+ standards, ensuring reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Scratch-resistant lenses enhance durability and longevity.
  • Polycarbonate lenses provide impact resistance for safety.
  • Clear lenses offer unobstructed vision.
  • Half-framed design for a lightweight feel.
  • Adjustable temples ensure a comfortable and secure fit for various head sizes.
  • Meets ANSI Z87.1+ standards for reliable eye protection.
  • Flexible nose bridge for added comfort.