Learn More
MSC Uvex Safety Glasses, Clear Lens, Scratch-Resistant, Black Frame
Catalog No. 5000507977
Encompass
Uvex Genesis XC safety glasses offer advanced protection and comfort. With a full-framed, wrap-around design and comfort brow, these glasses feature polycarbonate lenses with premium HydroShield Anti-Fog and Ultra-dura Hardcoat for enhanced visibility and durability.
  • Polycarbonate lens construction
  • Premium HydroShield Anti-Fog and Ultra-dura Hardcoat
  • Adjustable temples and flexible nose "fingers"
  • 9-base wrap-around lens for peripheral vision
  • Soft elastomer browguard for impact diffusion and comfort
  • Meets military Vo ballistic impact requirements
  • ANSI Z87.1+ - 2015 and CSA Z94.3 certified
  • U6 grade UV protection, 99.9% UVB/UVA filtration
  • Economical lens replacement and lifetime frame guarantee
  • Made in the USA
  • RX inserts available
Catalog No. 50-005-07977 Supplier MSC Supplier No. 06083794
Uvex Genesis XC safety glasses offer advanced protection and comfort. With a full-framed, wrap-around design and comfort brow, these glasses feature polycarbonate lenses with premium HydroShield Anti-Fog and Ultra-dura Hardcoat for enhanced visibility and durability.
  • Polycarbonate lens construction
  • Premium HydroShield Anti-Fog and Ultra-dura Hardcoat
  • Adjustable temples and flexible nose "fingers"
  • 9-base wrap-around lens for peripheral vision
  • Soft elastomer browguard for impact diffusion and comfort
  • Meets military Vo ballistic impact requirements
  • ANSI Z87.1+ - 2015 and CSA Z94.3 certified
  • U6 grade UV protection, 99.9% UVB/UVA filtration
  • Economical lens replacement and lifetime frame guarantee
  • Made in the USA
  • RX inserts available