Learn More
MSC Uvex Safety Glasses, Espresso Lens, Vapor Blue Frame, HydroShield Anti-Fog
Catalog No. 5000508749
Encompass
These safety glasses offer comprehensive eye protection with a half-frame design for comfort and durability. The espresso lens color and vapor blue frame provide a distinct appearance, while the HydroShield anti-fog coating ensures clear vision. They meet stringent safety standards, including ANSI Z87.1-2015 and CSA Z94.3-2007, making them ideal for general-purpose use.
  • Adjustable arm length for a customized fit.
  • Meets high impact standard ANSI Z87+.
  • Provides 99.9% UVA/UVB protection.
  • Universal eyewear size.
  • Polycarbonate lens material.
  • Complies with ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
  • HydroShield anti-fog lens coating.
Catalog No. 50-005-08749 Supplier MSC Supplier No. 32579419
These safety glasses offer comprehensive eye protection with a half-frame design for comfort and durability. The espresso lens color and vapor blue frame provide a distinct appearance, while the HydroShield anti-fog coating ensures clear vision. They meet stringent safety standards, including ANSI Z87.1-2015 and CSA Z94.3-2007, making them ideal for general-purpose use.
  • Adjustable arm length for a customized fit.
  • Meets high impact standard ANSI Z87+.
  • Provides 99.9% UVA/UVB protection.
  • Universal eyewear size.
  • Polycarbonate lens material.
  • Complies with ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
  • HydroShield anti-fog lens coating.