Learn More
MSC Uvex Safety Glasses, Genesis XC, Amber Lens, Black Frame, HydroShield Anti-Fog
Catalog No. 5000509276
Encompass
These safety glasses feature an amber polycarbonate lens with HydroShield Anti-Fog coating and a black half-frame. Designed for comfort and protection, they offer 99.9% UVA/UVB protection and meet ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
  • Amber lens color
  • Black half-frame design
  • Polycarbonate lens material
  • HydroShield Anti-Fog coating for enhanced fog-free performance and scratch-resistance
  • Hydrophilic and hydrophobic lens coating
  • Wraparound frame style
  • Adjustable temple
  • Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards
  • 99.9% UVA/UVB protection
Catalog No. 50-005-09276 Supplier MSC Supplier No. 32311532
These safety glasses feature an amber polycarbonate lens with HydroShield Anti-Fog coating and a black half-frame. Designed for comfort and protection, they offer 99.9% UVA/UVB protection and meet ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
  • Amber lens color
  • Black half-frame design
  • Polycarbonate lens material
  • HydroShield Anti-Fog coating for enhanced fog-free performance and scratch-resistance
  • Hydrophilic and hydrophobic lens coating
  • Wraparound frame style
  • Adjustable temple
  • Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards
  • 99.9% UVA/UVB protection