Learn More
MSC Uvex Safety Glasses, Hydroshield Anti-Fog, Clear Lens, ANSI Z87+, CSA Z94.3, Dielectric
Catalog No. 5000508228
Encompass
The Uvex Avatar Plus safety spectacles offer an optimal balance of technology and design, providing superior protection and comfort. Featuring a contoured shape that helps keep dust away, these spectacles combine a sporty look with a comfortable feel, enhancing worker well-being and productivity. Equipped with a Hydroshield anti-fog and anti-scratch lens coating, they meet ANSI Z87+ and CSA Z94.3 standards, ensuring reliable safety for various applications.
  • Contoured shape helps keep dust away
  • Hydroshield anti-fog and anti-scratch lens coating
  • Soft, flexible nose bridge and micro-textured temples ensure a comfortable and secure fit
  • Vented eyebrow pad enhances airflow
  • Meets ANSI Z87+ and CSA Z94.3 safety standards
  • Dielectric protection
  • Replaceable lenses available
Catalog No. 50-005-08228 Supplier MSC Supplier No. 65339855
The Uvex Avatar Plus safety spectacles offer an optimal balance of technology and design, providing superior protection and comfort. Featuring a contoured shape that helps keep dust away, these spectacles combine a sporty look with a comfortable feel, enhancing worker well-being and productivity. Equipped with a Hydroshield anti-fog and anti-scratch lens coating, they meet ANSI Z87+ and CSA Z94.3 standards, ensuring reliable safety for various applications.
  • Contoured shape helps keep dust away
  • Hydroshield anti-fog and anti-scratch lens coating
  • Soft, flexible nose bridge and micro-textured temples ensure a comfortable and secure fit
  • Vented eyebrow pad enhances airflow
  • Meets ANSI Z87+ and CSA Z94.3 safety standards
  • Dielectric protection
  • Replaceable lenses available