Learn More
MSC Uvex Safety Glasses, OTG, Clear Polycarbonate Lens, Anti-Fog, UV Protection
Catalog No. 5000508687
Encompass
Uvex Avatar Plus OTG Safety Glasses offer comfort and protection for demanding environments. They feature clear polycarbonate lenses with anti-fog, scratch-resistant, and UV protective coatings, designed to fit over prescription eyewear.
  • Anti-fog coatings improve visibility in humid conditions.
  • Scratch-resistant lens coating for durability.
  • Over-the-glasses (OTG) style fits over prescription eyewear.
  • Clear polycarbonate lens material for optical clarity.
  • UV protection safeguards eyes from harmful rays.
  • Universal size for a comfortable fit.
  • Flexible temple design enhances wearer comfort.
  • Floating nose pad for added comfort.
  • Meets ANSI Z87+ and CSA Z94.3 safety standards.
  • Nylon frame material for robust construction.
Catalog No. 50-005-08687 Supplier MSC Supplier No. 65340101
Uvex Avatar Plus OTG Safety Glasses offer comfort and protection for demanding environments. They feature clear polycarbonate lenses with anti-fog, scratch-resistant, and UV protective coatings, designed to fit over prescription eyewear.
  • Anti-fog coatings improve visibility in humid conditions.
  • Scratch-resistant lens coating for durability.
  • Over-the-glasses (OTG) style fits over prescription eyewear.
  • Clear polycarbonate lens material for optical clarity.
  • UV protection safeguards eyes from harmful rays.
  • Universal size for a comfortable fit.
  • Flexible temple design enhances wearer comfort.
  • Floating nose pad for added comfort.
  • Meets ANSI Z87+ and CSA Z94.3 safety standards.
  • Nylon frame material for robust construction.