Learn More
MSC Uvex Safety Glasses, Scratch-Resistant, Polycarbonate, Gray Lenses, Frameless
Catalog No. 5000508277
Encompass
These safety glasses feature scratch-resistant, polycarbonate, gray lenses with a frameless design. They include a black frame and adjustable temples, are part of the Protege series, and meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Frameless design
  • Wrap around frame
  • Gray lens color
  • Black frame color
  • Universal size
  • Protege series
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-005-08277 Supplier MSC Supplier No. 52882966
These safety glasses feature scratch-resistant, polycarbonate, gray lenses with a frameless design. They include a black frame and adjustable temples, are part of the Protege series, and meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Frameless design
  • Wrap around frame
  • Gray lens color
  • Black frame color
  • Universal size
  • Protege series
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards