Learn More
MSC Uvex Safety Glasses, SCT-Blue Lens, Anti-Fog, Half-Frame, Black
Catalog No. 5000508531
Encompass
The Uvex Genesis XC Safety Glasses offer high-performance protection and comfort. Featuring an SCT-Blue polycarbonate lens with HydroShield anti-fog and scratch-resistant coatings, these safety glasses are designed with a black half-frame and provide extensive coverage and adjustability for a secure fit.
  • Cushioned comfort and adjustability
  • Extended wrap-around lens for extra cheek and side coverage
  • Rx lens insert available
  • Soft, flexible nose fingers conform to various nasal profiles
  • 99.9% UVA/UVB protection
  • Polycarbonate lens and frame for durability
  • Lifetime frame guarantee and economical lens replacement system
Catalog No. 50-005-08531 Supplier MSC Supplier No. 32311466
The Uvex Genesis XC Safety Glasses offer high-performance protection and comfort. Featuring an SCT-Blue polycarbonate lens with HydroShield anti-fog and scratch-resistant coatings, these safety glasses are designed with a black half-frame and provide extensive coverage and adjustability for a secure fit.
  • Cushioned comfort and adjustability
  • Extended wrap-around lens for extra cheek and side coverage
  • Rx lens insert available
  • Soft, flexible nose fingers conform to various nasal profiles
  • 99.9% UVA/UVB protection
  • Polycarbonate lens and frame for durability
  • Lifetime frame guarantee and economical lens replacement system