Learn More
MSC Uvex Safety Glasses, SCT-Reflect 50, Black/Silver, ANSI Z87.1-2010
Catalog No. 5000509577
Encompass
These safety glasses offer exceptional comfort, protection, and style in a lightweight, sleek, frameless design. Made from polycarbonate, they provide 99.9% UV protection and meet ANSI Z87.1-2010 and CSA Z94.3 standards.
  • Dual nine-base, wrap-around unilens provides close-to-the-face coverage.
  • Adjustable nose piece for a personalized, comfortable fit.
  • Soft temple tip pads ensure a secure fit.
  • Lightweight, frameless design offers 180 degrees of unobstructed vision.
  • Unisex styling.
  • 99.9% ultra-violet protection.
Catalog No. 50-005-09577 Supplier MSC Supplier No. 51469005
These safety glasses offer exceptional comfort, protection, and style in a lightweight, sleek, frameless design. Made from polycarbonate, they provide 99.9% UV protection and meet ANSI Z87.1-2010 and CSA Z94.3 standards.
  • Dual nine-base, wrap-around unilens provides close-to-the-face coverage.
  • Adjustable nose piece for a personalized, comfortable fit.
  • Soft temple tip pads ensure a secure fit.
  • Lightweight, frameless design offers 180 degrees of unobstructed vision.
  • Unisex styling.
  • 99.9% ultra-violet protection.