Learn More
MSC Uvex Safety Goggles, Anti-Fog, Gray Polycarbonate Lenses, Indirect Vent
Catalog No. 5000508182
Encompass
These safety goggles feature indirect ventilation and a gray frame, designed for universal fit. They offer anti-fog gray polycarbonate lenses for improved visibility, making them suitable for various industrial and laboratory environments requiring eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Tinted lenses.
  • Indirect ventilation.
  • Provides protection against dust and splash.
  • Gray polycarbonate lenses.
  • Gray elastomer frame.
  • Universal size.
  • UV protection.
  • Meets ANSI Z87.1+ standards.
  • Includes headband made of neoprene.
Catalog No. 50-005-08182 Supplier MSC Supplier No. 53160123
These safety goggles feature indirect ventilation and a gray frame, designed for universal fit. They offer anti-fog gray polycarbonate lenses for improved visibility, making them suitable for various industrial and laboratory environments requiring eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Tinted lenses.
  • Indirect ventilation.
  • Provides protection against dust and splash.
  • Gray polycarbonate lenses.
  • Gray elastomer frame.
  • Universal size.
  • UV protection.
  • Meets ANSI Z87.1+ standards.
  • Includes headband made of neoprene.