Learn More
MSC Uvex Safety Goggles, Indirect Vent, Clear Polycarbonate Lens, Anti-Fog
Catalog No. 5000508396
Encompass
Uvex Strategy Series Safety Goggles offer comfortable and reliable eye protection. Designed for demanding environments, they feature clear polycarbonate lenses with Uvextra AF anti-fog coating for enhanced visibility and indirect ventilation for fine particle protection. These goggles are also designed to fit over most prescription eyewear.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Scratch-resistant clear polycarbonate lenses offer durability.
  • Indirect ventilation with foam lining provides protection against fine particles, dust, and splash.
  • Designed to fit comfortably over most prescription eyewear.
  • Provides 99.9% UV protection.
  • Lightweight thermoplastic elastomer body ensures a snug and comfortable fit.
  • Fabric headband for secure wear.
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards.
Catalog No. 50-005-08396 Supplier MSC Supplier No. 51468874
Uvex Strategy Series Safety Goggles offer comfortable and reliable eye protection. Designed for demanding environments, they feature clear polycarbonate lenses with Uvextra AF anti-fog coating for enhanced visibility and indirect ventilation for fine particle protection. These goggles are also designed to fit over most prescription eyewear.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Scratch-resistant clear polycarbonate lenses offer durability.
  • Indirect ventilation with foam lining provides protection against fine particles, dust, and splash.
  • Designed to fit comfortably over most prescription eyewear.
  • Provides 99.9% UV protection.
  • Lightweight thermoplastic elastomer body ensures a snug and comfortable fit.
  • Fabric headband for secure wear.
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards.