Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Catalog No. 19351451
Change view
Click to view available options
Frame Color:
Black
Metallic Blue
Lens Coating:
Anti-fog
Hardcoat
Scratch-resistant
Lens Tint:
Amber
Clear
Espresso
SCT-Reflect 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-351-451 Black Scratch-resistant Clear
19-185-650 Black Anti-fog Clear
19-351-460 Metallic Blue Anti-fog SCT-Reflect 50
19-351-455 Black Anti-fog Amber
19-351-457 Metallic Blue Hardcoat Clear
19-351-456 Black Anti-fog SCT-Reflect 50
19-351-454 Black Anti-fog Espresso
19-351-452 Black Anti-fog Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 19-351-451 Supplier Honeywell™ Supplier No. S0600
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Eye protection against impact, sun, wind, dust and airborne debris
  • For dirty, high-particulate work environments
  • With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
  • Soft and flexible nasal pads
  • Soft, high-quality face cushioning and padded temple tips
  • Highly adjustable flame-resistant headband
  • Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-2003.
Lens Tint Clear
For Use With (Application) For applications like grinding, chipping, shaving and sanding
Frame Color Black
Temple Style Removable