Learn More
MSC Uvex Seismic Sealed Safety Glasses, Polycarbonate Lens, Clear, Black Frame, HydroShield Anti-Fog
Catalog No. 5000509292
Encompass
Uvex Seismic Sealed Safety Glasses with HydroShield Anti-Fog coating are designed for demanding environments, offering comprehensive protection against impact, wind, dust, and debris. They feature a clear polycarbonate lens and a black frame, combining safety with comfort through soft nasal pads and a highly adjustable, flame-resistant headband.
  • Offers protection against impact, wind, dust, and debris.
  • Designed for high-particulate environments.
  • Features a clear polycarbonate lens with HydroShield anti-fog and scratch-resistant coating.
  • Black, full-framed design.
  • Cushioned-lined subframe and removable headband for comfort.
  • Soft, flexible nasal pads and face cushioning.
  • Flame-resistant, adjustable headband.
  • Provides UV protection.
  • Meets ANSI Z87.1+ standards.
  • Universal size for various facial profiles.
Catalog No. 50-005-09292 Supplier MSC Supplier No. 39652185
Uvex Seismic Sealed Safety Glasses with HydroShield Anti-Fog coating are designed for demanding environments, offering comprehensive protection against impact, wind, dust, and debris. They feature a clear polycarbonate lens and a black frame, combining safety with comfort through soft nasal pads and a highly adjustable, flame-resistant headband.
  • Offers protection against impact, wind, dust, and debris.
  • Designed for high-particulate environments.
  • Features a clear polycarbonate lens with HydroShield anti-fog and scratch-resistant coating.
  • Black, full-framed design.
  • Cushioned-lined subframe and removable headband for comfort.
  • Soft, flexible nasal pads and face cushioning.
  • Flame-resistant, adjustable headband.
  • Provides UV protection.
  • Meets ANSI Z87.1+ standards.
  • Universal size for various facial profiles.