Learn More
Honeywell Safety Products™ Uvex™ Skyper™ Safety Glasses
Catalog No. 19038311
Change view
Click to view available options
Lens Tint:
Clear
IR Filter Shade 2.0
IR Filter Shade 3.0
IR Filter Shade 5.0
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-038-311 IR Filter Shade 5.0
19-038-310 IR Filter Shade 3.0
19-038-309 IR Filter Shade 2.0
19-037-656 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-038-311 Supplier Honeywell Safety Products™ Supplier No. S1908
This item is not returnable. View return policy
Custom fit with superior peripheral protection
  • Polycarbonate lenses with single-lens design feature Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic coating
  • Wraparound, impact resistant lenses block 99.9% of harmful UVA and UVB radiation
  • Black frames with molded-in browguard reduce the danger of flying particulates
  • Molded-in nosebridge offers comfort and safety in a variety of work applications
  • Patented, adjustable, comfort-cushioned Duoflex™ temples
  • Simple lens replacement system, ratcheting lens inclination
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-1999 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Lens Coating Scratch-resistant
Description Shade 5.0 Infra-dura
Lens Tint IR Filter Shade 5.0
Frame Color Black
Lens Material Polycarbonate