Learn More
Honeywell Safety Products™ Uvex™ Stealth™ Chemical-Splash Goggles
Catalog No. 189992315
Change view
Click to view available options
Frame Color:
Light Gray, Charcoal
Teal, Light Gray
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Amber
Clear
Lens Type:
Durastreme
Uvextreme Anti-fog
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint Lens Type
18-999-2315 Teal, Light Gray Anti-fog Clear Uvextreme Anti-fog
18-999-2313 Light Gray, Charcoal Anti-fog Amber Uvextreme Anti-fog
18-999-2312 Light Gray, Charcoal Anti-fog, Scratch-resistant Clear Durastreme
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-999-2315 Supplier Honeywell Safety Products™ Supplier No. S39610C
This item is not returnable. View return policy
Sleek, low-profile design fits well with respirators, hardhats or welding helmets
  • Polycarbonate lens is coated with uvextreme™ anti-fog coating (XTR) for protection against fogging, scratches, static dust, and 99.9% of harmful UV rays up to 385nm
  • Toric lens provides superior optics and improves peripheral vision
  • Combines durable polypropylene frame and soft, flexible elastomer inner flange
  • Provides superior splash protection
  • Indirect ventilation system directs airflow over the lens to minimize fogging and improve vision—without compromising splash and impact protection
  • Neoprene headband is textured to prevent slipping and features quick-adjust clips for easy and secure fit
  • Snap-off/snap-on lens replacement system suits many applications and environments
Order Info
Visit www.fishersci.com (keyword: uvex stealth) for replacement lenses and Rx carrier adapter that allows prescription glasses to be used with goggles.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type Uvextreme Anti-fog
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2010
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Teal, Light Gray
Headband Material Neoprene
No. per Case 12/Cs.
Ventilation Details Indirect ventilation system
Material Polypropylene (Frame)
Lens Material Polycarbonate
Frame Material Polypropylene